ADVH14DBBB05 AChemBlock

tert-Butyl 1-aminocyclopropanecarboxylate

  • MDL number MFCD21143671
  • Cas Number : 159871-51-9
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-aminocyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40868
Availability details for
Quantity
  • Availability
  • Add to Cart