ADVH14DBBB0C AChemBlock

Methyl a-(Aminomethyl)benzeneacetate hydrochloride

  • MDL number MFCD06740372
  • Cas Number : 91012-17-8
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-a-aminomethyl-benzeneacetate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35205
Availability details for
Quantity
  • Availability
  • Add to Cart