ADVH14DBBB1A AChemBlock

(R)-Methyl 2-amino-2-(4-bromophenyl)acetate hydrochloride

  • MDL number MFCD12910952
  • Cas Number : 1391389-21-1
  • Molecular Weight : 280.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-methyl-2-amino-2-4-bromophenyl-acetate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41417
Availability details for
Quantity
  • Availability
  • Add to Cart