ADVH14DBBB28 AChemBlock

5-Bromo-N-Fmoc-DL-tryptophan

  • MDL number MFCD02682357
  • Cas Number : 925939-82-8
  • Molecular Weight : 505.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-n-fmoc-dl-tryptophan.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35065
Availability details for
Quantity
  • Availability
  • Add to Cart