ADVH14DBBB59 AChemBlock

4-(Fmoc-NH2)-1-(2,4-F2-benzyl)-4-carboxypiperidine

  • MDL number MFCD08751586
  • Cas Number : 916422-05-4
  • Molecular Weight : 492.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fmoc-nh2-1-2-4-f2-benzyl-4-carboxypiperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35110
Availability details for
Quantity
  • Availability
  • Add to Cart