ADVH14DBBB6A AChemBlock

N-Boc-N-(2-methylpropyl)-glycine

  • MDL number MFCD17286086
  • Cas Number : 194206-80-9
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-n-2-methylpropyl-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40126
Availability details for
Quantity
  • Availability
  • Add to Cart