ADVH14DBBB71 AChemBlock

N-[(4-Chlorophenyl)methyl]-N-Boc-glycine

  • MDL number MFCD11964025
  • Cas Number : 1182070-86-5
  • Molecular Weight : 299.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-chlorophenyl-methyl-n-boc-glycine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42435
Availability details for
Quantity
  • Availability
  • Add to Cart