ADVH14DBBBAB AChemBlock

3-[(Fmoc-amino)methyl]benzoic acid

  • MDL number MFCD00273463
  • Cas Number : 155369-11-2
  • Molecular Weight : 373.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fmoc-amino-methyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40961
Availability details for
Quantity
  • Availability
  • Add to Cart