ADVH14DBBBC0 AChemBlock

Methyl 3-amino-3-(4-fluorophenyl)propanoate hydrochloride

  • MDL number MFCD13196082
  • Cas Number : 181519-32-4
  • Molecular Weight : 233.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-3-4-fluorophenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40370
Availability details for
Quantity
  • Availability
  • Add to Cart