ADVH14DBBBED AChemBlock

3-(Trifluoromethoxy)-D-phenylalanine

  • MDL number MFCD07371972
  • Cas Number : 1241681-55-9
  • Molecular Weight : 249.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-trifluoromethoxy-d-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42100
Availability details for
Quantity
  • Availability
  • Add to Cart