ADVH14DBBBF0 AChemBlock

a-(Methylamino)benzeneacetic acid hydrochloride

  • MDL number MFCD06795795
  • Cas Number : 28544-42-5
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-methylamino-benzeneacetic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39058
Availability details for
Quantity
  • Availability
  • Add to Cart