ADVH14DBBC06 AChemBlock

4-(2-Methylphenyl)-L-phenylalanine

  • MDL number MFCD17215561
  • Cas Number : 758685-64-2
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-2-methylphenyl-l-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36223
Availability details for
Quantity
  • Availability
  • Add to Cart