ADVH14DBBC12 AChemBlock

(S)-3-Amino-4-methylpentanoic acid hydrochloride

  • MDL number MFCD06202413
  • Cas Number : 402587-64-8
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-4-methylpentanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38250
Availability details for
Quantity
  • Availability
  • Add to Cart