ADVH14DBBC44 AChemBlock

N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride

  • MDL number MFCD24842520
  • Cas Number : 1428330-91-9
  • Molecular Weight : 264.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n6-2-propyn-1-yloxy-carbonyl-l-lysine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41280
Availability details for
Quantity
  • Availability
  • Add to Cart