ADVH14DBBC69 AChemBlock

a-Methyl-1-naphthaleneacetic acid

  • MDL number MFCD02066245
  • Cas Number : 3117-51-9
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-methyl-1-naphthaleneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38898
Availability details for
Quantity
  • Availability
  • Add to Cart