ADVH14DBBC70 AChemBlock

Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate

  • MDL number MFCD07779434
  • Cas Number : 51656-91-8
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-4-dioxaspiro-4-5-dec-8-ylideneacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37698
Availability details for
Quantity
  • Availability
  • Add to Cart