ADVH14DBBC88 AChemBlock

2-(4-Aminophenoxy)-2-methyl-propanoic acid hydrochloride

  • MDL number MFCD12022380
  • Cas Number : 117011-70-8
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-aminophenoxy-2-methyl-propanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42478
Availability details for
Quantity
  • Availability
  • Add to Cart