ADVH14DBBC8D AChemBlock

1-Carboxy-(2-trifluoromethylphenyl)cyclopropane

  • MDL number MFCD07374436
  • Cas Number : 886366-06-9
  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-carboxy-2-trifluoromethylphenyl-cyclopropane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35422
Availability details for
Quantity
  • Availability
  • Add to Cart