ADVH14DBBC93 AChemBlock

1-(4-Ethoxyphenyl)cyclohexanecarboxylic acid

  • MDL number MFCD11541475
  • Cas Number : 36263-67-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-ethoxyphenyl-cyclohexanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38528
Availability details for
Quantity
  • Availability
  • Add to Cart