ADVH14DBBC9B AChemBlock

2-Fluoro-biphenyl-4-acetic acid

  • MDL number MFCD09840622
  • Cas Number : 5001-96-7
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-biphenyl-4-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37865
Availability details for
Quantity
  • Availability
  • Add to Cart