ADVH14DBBCC2 AChemBlock

2-Chloro-N-(4-pyridinylmethyl)-acetamide

  • MDL number MFCD02807834
  • Cas Number : 125794-21-0
  • Molecular Weight : 184.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-n-4-pyridinylmethyl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42031
Availability details for
Quantity
  • Availability
  • Add to Cart