ADVH14DBBD1E AChemBlock

4-[3-(N,N-Dimethylamino)propoxy]aniline

  • MDL number MFCD08687611
  • Cas Number : 62424-88-8
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-3-n-n-dimethylamino-propoxy-aniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37001
Availability details for
Quantity
  • Availability
  • Add to Cart