ADVH14DBBD6F AChemBlock

1-Boc-4-cyano-4-(4-methylphenyl)-piperidine

  • MDL number MFCD11227119
  • Cas Number : 906369-93-5
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-cyano-4-4-methylphenyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35235
Availability details for
Quantity
  • Availability
  • Add to Cart