ADVH14DBBD9C AChemBlock

1-Boc-3-phenylamino-piperidine

  • MDL number MFCD07786036
  • Cas Number : 183207-67-2
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-phenylamino-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40331
Availability details for
Quantity
  • Availability
  • Add to Cart