ADVH14DBBDB0 AChemBlock

1-Boc-4-(4-chlorophenyl)-4-hydroxypiperidine

  • MDL number MFCD09953408
  • Cas Number : 235109-63-4
  • Molecular Weight : 311.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-4-chlorophenyl-4-hydroxypiperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39490
Availability details for
Quantity
  • Availability
  • Add to Cart