ADVH14DBBDBB AChemBlock

N-Cyclopropyl-N-methyl-4-piperidinamine dihydrochloride

  • MDL number MFCD15142893
  • Cas Number : 1048919-83-0
  • Molecular Weight : 227.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-cyclopropyl-n-methyl-4-piperidinamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42836
Availability details for
Quantity
  • Availability
  • Add to Cart