ADVH14DBBDC2 AChemBlock

1-Ethyl-3-piperidinamine dihydrochloride

  • MDL number MFCD19982401
  • Cas Number : 6789-94-2
  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-ethyl-3-piperidinamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36640
Availability details for
Quantity
  • Availability
  • Add to Cart