ADVH14DBBDF1 AChemBlock

4-Oxo-1-(oxan-4-yl)cyclohexanecarbonitrile

  • MDL number MFCD17013955
  • Cas Number : 1202462-46-1
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-oxo-1-oxan-4-yl-cyclohexanecarbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42327
Availability details for
Quantity
  • Availability
  • Add to Cart