ADVH14DBBDFA AChemBlock

5-(4-Morpholinylmethyl)-2-thiazolamine

  • MDL number MFCD19254592
  • Cas Number : 3008-62-6
  • Molecular Weight : 199.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-4-morpholinylmethyl-2-thiazolamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38962
Availability details for
Quantity
  • Availability
  • Add to Cart