ADVH14DBBE33 AChemBlock

1-Cbz-3-(aminomethyl)-pyrrolidine

  • MDL number MFCD06858456
  • Cas Number : 315717-77-2
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-cbz-3-aminomethyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38879
Availability details for
Quantity
  • Availability
  • Add to Cart