ADVH14DBBE4C AChemBlock

3-(Boc-amino)-1-pyrrolidineacetic acid

  • MDL number MFCD01861775
  • Cas Number : 261715-71-3
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-boc-amino-1-pyrrolidineacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39262
Availability details for
Quantity
  • Availability
  • Add to Cart