ADVH14DBBE7E AChemBlock

(R)-1-Fmoc-3-pyrrolidinecarboxylic acid

  • MDL number MFCD14635841
  • Cas Number : 193693-65-1
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-fmoc-3-pyrrolidinecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40137
Availability details for
Quantity
  • Availability
  • Add to Cart