ADVH14DBBE84 AChemBlock

3-(2-Oxo-1-pyrrolidinyl)-benzoic acid

  • MDL number MFCD01422242
  • Cas Number : 515813-05-5
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-oxo-1-pyrrolidinyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37708
Availability details for
Quantity
  • Availability
  • Add to Cart