ADVH14DBBE89 AChemBlock

3-Isopropoxybenzaldehyde

  • MDL number MFCD01922005
  • Cas Number : 75792-33-5
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-isopropoxybenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36227
Availability details for
Quantity
  • Availability
  • Add to Cart