ADVH14DBBEA8 AChemBlock

Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate

  • MDL number MFCD09759206
  • Cas Number : 3697-68-5
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-hydroxymethyl-cyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38485
Availability details for
Quantity
  • Availability
  • Add to Cart