ADVH14DBBEB4 AChemBlock

1-(3-Azetidinyl)-4-methylpiperazine trihydrochloride

  • MDL number MFCD28118247
  • Cas Number : 302355-82-4
  • Molecular Weight : 264.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-azetidinyl-4-methylpiperazine-trihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38949
Availability details for
Quantity
  • Availability
  • Add to Cart