ADVH14DBBECE AChemBlock

4-(Azetidin-3-yloxy)-benzonitrile hydrochloride

  • MDL number MFCD26407821
  • Cas Number : 949100-15-6
  • Molecular Weight : 210.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-azetidin-3-yloxy-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34874
Availability details for
Quantity
  • Availability
  • Add to Cart