ADVH14DBBEE3 AChemBlock

2'-[(Boc-amino)methyl]-biphenyl-2-carboxylic acid

  • MDL number MFCD04974227
  • Cas Number : 158066-11-6
  • Molecular Weight : 327.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-boc-amino-methyl-biphenyl-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40908
Availability details for
Quantity
  • Availability
  • Add to Cart