ADVH14DBBF0B AChemBlock

3-[(2-Bromophenyl)methyl]azetidine hydrochloride

  • MDL number MFCD31536908
  • Cas Number : 1016772-34-1
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-bromophenyl-methyl-azetidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43052
Availability details for
Quantity
  • Availability
  • Add to Cart