ADVH14DBBF16 AChemBlock

1-Boc-2-ethyl-2-azetidinecarboxylic acid

  • MDL number MFCD12406957
  • Cas Number : 610791-07-6
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-2-ethyl-2-azetidinecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37094
Availability details for
Quantity
  • Availability
  • Add to Cart