ADVH14DBBF20 AChemBlock

N-(3-Azetidinylmethyl)-acetamide hydrochloride

  • MDL number MFCD11974383
  • Cas Number : 124668-51-5
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-3-azetidinylmethyl-acetamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42074
Availability details for
Quantity
  • Availability
  • Add to Cart