ADVH14DBBFC2 AChemBlock

4-(Boc-amino)-3-oxobutanoic acid ethyl ester

  • MDL number MFCD11616704
  • Cas Number : 67706-68-7
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-boc-amino-3-oxobutanoic-acid-ethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36665
Availability details for
Quantity
  • Availability
  • Add to Cart