ADVH14DBBFF2 AChemBlock

a,g-Dioxo-cyclopropanebutanoic acid ethyl ester

  • MDL number MFCD09052462
  • Cas Number : 21080-80-8
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-g-dioxo-cyclopropanebutanoic-acid-ethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39812
Availability details for
Quantity
  • Availability
  • Add to Cart