ADVH14DBC00B AChemBlock

3,5-Dimethoxy-b-oxo-benzenepentanenitrile

  • MDL number MFCD22199282
  • Cas Number : 1000895-54-4
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-5-dimethoxy-b-oxo-benzenepentanenitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43138
Availability details for
Quantity
  • Availability
  • Add to Cart