ADVH14DBC055 AChemBlock

5-Fluoro-2-methoxypyridine

  • MDL number MFCD03095252
  • Cas Number : 51173-04-7
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-2-methoxypyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37735
Availability details for
Quantity
  • Availability
  • Add to Cart