ADVH14DBC05A AChemBlock

4-Chloro-2,6-dimethyl-pyridine

  • MDL number MFCD01646174
  • Cas Number : 3512-75-2
  • Molecular Weight : 141.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-2-6-dimethyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38625
Availability details for
Quantity
  • Availability
  • Add to Cart