ADVH14DBC065 AChemBlock

6-Chloro-3-pyridineethanamine

  • MDL number MFCD08449285
  • Cas Number : 54127-64-9
  • Molecular Weight : 156.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3-pyridineethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37543
Availability details for
Quantity
  • Availability
  • Add to Cart