ADVH14DBC06C AChemBlock

3-Methyl-2-pyridinemethanamine

  • MDL number MFCD04116209
  • Cas Number : 153936-26-6
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-2-pyridinemethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40986
Availability details for
Quantity
  • Availability
  • Add to Cart