ADVH14DBC091 AChemBlock

6-(Methoxycarbonyl)picolinic acid

  • MDL number MFCD00806109
  • Cas Number : 7170-36-7
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-methoxycarbonyl-picolinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36447
Availability details for
Quantity
  • Availability
  • Add to Cart