ADVH14DBC11A AChemBlock

4-Hydroxy-4-(4-methylphenyl)piperidine hydrochloride

  • MDL number MFCD28108385
  • Cas Number : 57988-60-0
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxy-4-4-methylphenyl-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37270
Availability details for
Quantity
  • Availability
  • Add to Cart